siRNA Details
| Gene Symbol: UBL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048241.2 | NP_001041706.1 | 3 | 247 | ||
| NM_024292.3 | NP_077268.1 | 3 | 315 | |||
| GenBank mRNA | AF313915.1 | - | 3 | 196 | ||
| AK128885.1 | - | 3 | 315 | |||
| AK311889.1 | - | 3 | 235 | |||
| BC007053.1 | - | 3 | 269 | 3827384 | ||
| BC110839.1 | - | 3 | 163 | 6474495 | ||
| BP213482.1 | - | 3 | 235 | |||
| BT007355.1 | - | 2 | 131 | |||

UBL5
Genomic Map