siRNA Details
| Gene Symbol: LSM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016199.2 | NP_057283.1 | 3 | 158 | ||
| XM_017026869.1 | XP_016882358.1 | 4 | 301 | |||
| GenBank mRNA | AF182293.1 | - | 2 | 118 | ||
| BC018621.1 | - | 2 | 111 | 4855162 | ||
| BI553911.1 | - | 3 | 162 | |||

LSM7
Genomic Map