siRNA Details
| Gene Symbol: GLRX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016417.2 | NP_057501.2 | 2 | 634 | ||
| GenBank mRNA | AB223038.1 | - | 2 | 573 | ||
| AF113691.1 | - | 204 | ||||
| BC023528.2 | - | 2 | 584 | 3613760 | ||
| BC047680.1 | - | 2 | 580 | 6066312 | ||
| BC105275.1 | - | 1 | 111 | |||
| BI834047.1 | - | 2 | 382 | |||
| BX248075.1 | - | 2 | 587 | |||

GLRX5
Genomic Map