siRNA Details
| Gene Symbol: HMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018942.2 | NP_061815.2 | 2 | 823 | ||
| GenBank mRNA | M99587.1 | - | 2 | 810 | ||
| Gene Symbol: HMX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105574.1 | NP_001099044.1 | 2 | 692 | ||

HMX1
Genomic Map