siRNA Details
| Gene Symbol: HOXC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018953.3 | NP_061826.1 | 2 | 581 | ||
| NR_003084.2 | 2 | 615 | ||||
| GenBank mRNA | BC034922.1 | - | 2 | 357 | ||
| BC044635.1 | - | 2 | 357 | |||
| BM786543.1 | - | 2 | 494 | |||
| CN309383.1 | - | 2 | 157 | |||

HOXC5
Genomic Map