siRNA Details
| Gene Symbol: LRFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024036.4 | NP_076941.2 | 1 | 1662 | ||
| XM_005274239.3 | XP_005274296.1 | 2 | 1542 | |||
| GenBank mRNA | AB209005.1 | - | 1 | 1283 | ||
| BC000207.1 | - | 1 | 399 | |||
| BC014040.2 | - | 1 | 406 | |||
| BC015581.2 | - | 1 | 1565 | 4637053 | ||
| BC027475.1 | - | 1 | 1001 | |||
| BC094813.1 | - | 1 | 740 | |||

LRFN4
Genomic Map