siRNA Details
| Gene Symbol: KLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016270.3 | NP_057354.1 | 3 | 1490 | ||
| GenBank mRNA | AF134053.1 | - | 3 | 1392 | ||
| AF205849.1 | - | 3 | 1476 | |||
| AK225820.1 | - | 3 | 1489 | |||
| BC071983.1 | - | 3 | 1479 | 5480246 | ||
| BQ581981.1 | - | 2 | 526 | |||

KLF2
Genomic Map