siRNA Details
| Gene Symbol: ZNF358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018083.4 | NP_060553.4 | 2 | 1919 | ||
| XM_005272460.3 | XP_005272517.1 | 2 | 1908 | |||
| XM_011527695.1 | XP_011525997.1 | 2 | 1810 | |||
| XM_017026296.1 | XP_016881785.1 | 2 | 1913 | |||
| GenBank mRNA | AK001252.2 | - | 1 | 1697 | ||
| BC003585.1 | - | 1 | 1150 | |||
| BC014002.1 | - | 1 | 1610 | 3925722 | ||
| C15772.1 | - | 1 | 330 | |||

ZNF358
Genomic Map