siRNA Details
| Gene Symbol: BEX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018476.3 | NP_060946.3 | 3 | 803 | ||
| GenBank mRNA | AF183416.1 | - | 1 | 565 | ||
| AF220189.1 | - | 3 | 729 | |||
| AF237783.1 | - | 3 | 734 | |||
| AY833561.1 | - | 3 | 725 | |||
| BC018615.2 | - | 1 | 569 | |||
| BM465948.1 | - | 3 | 744 | |||

BEX1
Genomic Map