siRNA Details
| Gene Symbol: EXOSC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019037.2 | NP_061910.1 | 3 | 527 | ||
| XM_011517134.2 | XP_011515436.1 | 3 | 385 | |||
| GenBank mRNA | AF281133.1 | - | 3 | 520 | ||
| AK000598.1 | - | 3 | 444 | |||
| BC002777.2 | - | 3 | 441 | 3613853 | ||
| BM839994.1 | - | 3 | 527 | |||

EXOSC4
Genomic Map