siRNA Details
| Gene Symbol: CABP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019855.4 | NP_062829.1 | 3 | 246 | ||
| XM_017026992.1 | XP_016882481.1 | 5 | 447 | |||
| GenBank mRNA | AF169159.1 | - | 3 | 239 | ||
| BC126133.1 | - | 3 | 154 | 8991849 | ||
| BC126135.1 | - | 3 | 154 | 8991851 | ||

CABP5
Genomic Map