siRNA Details
| Gene Symbol: RETN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193374.1 | NP_001180303.1 | 4 | 438 | ||
| NM_020415.3 | NP_065148.1 | 4 | 447 | |||
| GenBank mRNA | AF205952.1 | - | 3 | 425 | ||
| AF290874.1 | - | 4 | 443 | |||
| AF323081.1 | - | 4 | 444 | |||
| AY207314.1 | - | 4 | 438 | |||
| AY359066.1 | - | 3 | 425 | |||

RETN
Genomic Map