siRNA Details
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250612.3 | XP_005250669.1 | 1 | 832 | ||
XM_005250613.4 | XP_005250670.2 | 2 | 424 | |||
XM_005250614.2 | XP_005250671.1 | 2 | 424 | |||
XM_006716140.3 | XP_006716203.1 | 2 | 424 | |||
XM_006716141.3 | XP_006716204.1 | 2 | 424 | |||
XM_017012885.1 | XP_016868374.1 | 2 | 424 | |||
XM_017012886.1 | XP_016868375.1 | 2 | 424 |
Gene Symbol: UPK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030570.3 | NP_085047.1 | 1 | 511 | ||
NM_182683.2 | NP_872624.1 | 2 | 346 | |||
NM_182684.2 | NP_872625.1 | 2 | 346 | |||
GenBank mRNA | AK056760.1 | - | 2 | 154 | ||
AK290096.1 | - | 1 | 414 | |||
AY233462.1 | - | 2 | 179 | |||
BC004304.1 | - | 1 | 335 | 3625247 | ||
BE615406.1 | - | 2 | 345 | |||
BM921749.1 | - | 370 | ||||
BQ689127.1 | - | 341 | ||||
BU150542.1 | - | 341 |
