siRNA Details
| Gene Symbol: ALKBH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032306.3 | NP_115682.1 | 4 | 930 | ||
| XM_005259658.4 | XP_005259715.1 | 4 | 506 | |||
| XM_017027355.1 | XP_016882844.1 | 3 | 787 | |||
| GenBank mRNA | AK223336.1 | - | 4 | 553 | ||
| AK311956.1 | - | 4 | 569 | |||
| AY358858.1 | - | 4 | 930 | |||
| AY427650.2 | - | 4 | 930 | |||
| BC004393.1 | - | 4 | 585 | 3635540 | ||

ALKBH7
Genomic Map