siRNA Details
| Gene Symbol: NICN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032316.3 | NP_115692.1 | 6 | 2223 | ||
| GenBank mRNA | AK094248.1 | - | 7 | 1928 | ||
| AK301846.1 | - | 6 | 1429 | |||
| BC005050.1 | - | 7 | 1156 | 2820022 | ||
| BC050653.1 | - | 7 | 1531 | 6015080 | ||

NICN1
Genomic Map