siRNA Details
Gene Symbol: LINC00152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024204.1 | 4 | 764 | |||
NR_024205.1 | 3 | 600 | ||||
NR_024206.1 | 2 | 454 | ||||
GenBank mRNA | BC009508.2 | - | 2 | 419 | ||
BC070202.1 | - | 377 | ||||
BM989746.1 | - | 2 | 449 | |||
CF129206.1 | - | 2 | 492 |
