siRNA Details
| Gene Symbol: MUCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058173.2 | NP_477521.1 | 2 | 158 | ||
| GenBank mRNA | AF414087.1 | - | 2 | 136 | ||
| AI905624.1 | - | 2 | 159 | |||
| AY359062.1 | - | 2 | 129 | |||
| AY651259.1 | - | 2 | 90 | |||
| BC111421.1 | - | 2 | 123 | 40021582 | ||

MUCL1
Genomic Map