siRNA Details
| Gene Symbol: GLI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138465.3 | NP_612474.1 | 3 | 274 | ||
| GenBank mRNA | AB209654.1 | - | 3 | 5261 | ||
| AY129020.1 | - | 1 | 170 | |||
| BC014165.2 | - | 2 | 240 | 4549240 | ||
| BI761835.1 | - | 3 | 274 | |||
| Gene Symbol: ZFP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL831951.1 | - | 3 | 2356 | ||

GLI4
Genomic Map