siRNA Details
| Gene Symbol: TECR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321170.1 | NP_001308099.1 | 12 | 884 | ||
| NM_138501.5 | NP_612510.1 | 11 | 810 | |||
| NR_038103.1 | 11 | 842 | ||||
| NR_038104.1 | 11 | 875 | ||||
| XM_006722945.1 | XP_006723008.1 | 12 | 928 | |||
| XM_011528442.2 | XP_011526744.1 | 11 | 758 | |||
| GenBank mRNA | AF038958.1 | - | 11 | 756 | ||
| AF038959.1 | - | 6 | 239 | |||
| AF222742.1 | - | 11 | 731 | |||
| AK001416.1 | - | 11 | 774 | |||
| AK094087.1 | - | 12 | 851 | |||
| AK299134.1 | - | 12 | 848 | |||
| AK315412.1 | - | 11 | 774 | |||
| BC000174.2 | - | 10 | 670 | |||
| BC002425.1 | - | 11 | 742 | 3163592 | ||
| BC005952.1 | - | 11 | 753 | 4249841 | ||
| BC007801.2 | - | 11 | 760 | 4128702 | ||
| BC013881.2 | - | 11 | 758 | 3928698 | ||
| BT007179.1 | - | 10 | 680 | |||
| CR456892.1 | - | 10 | 680 | |||

TECR
Genomic Map