siRNA Details
| Gene Symbol: TMEM134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001078650.2 | NP_001072118.1 | 6 | 606 | ||
| NM_001078651.2 | NP_001072119.1 | 7 | 624 | |||
| NM_025124.3 | NP_079400.1 | 7 | 651 | |||
| NR_073409.1 | 7 | 818 | ||||
| NR_073410.1 | 6 | 590 | ||||
| NR_073411.1 | 7 | 662 | ||||
| NR_073412.1 | 7 | 783 | ||||
| NR_073413.1 | 7 | 635 | ||||
| XM_006718693.2 | XP_006718756.1 | 6 | 574 | |||
| XM_017018356.1 | XP_016873845.1 | 6 | 752 | |||
| GenBank mRNA | AK025402.1 | - | 7 | 667 | ||
| BC013883.1 | - | 7 | 787 | 3678954 | ||
| BC125134.1 | - | 6 | 574 | 40119367 | ||
| BG488634.1 | - | 7 | 573 | |||
| BG719036.1 | - | 6 | 586 | |||
| BM920758.1 | - | 619 | ||||
| BX400047.2 | - | 7 | 739 | |||
| CN336445.1 | - | 7 | 485 | |||
| DN999439.1 | - | 6 | 595 | |||

TMEM134
Genomic Map