siRNA Details
| Gene Symbol: UPK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030570.3 | NP_085047.1 | 3 | 907 | ||
| NM_182683.2 | NP_872624.1 | 4 | 742 | |||
| NM_182684.2 | NP_872625.1 | 5 | 822 | |||
| GenBank mRNA | AK056760.1 | - | 4 | 550 | ||
| AK290096.1 | - | 3 | 810 | |||
| AY233462.1 | - | 5 | 655 | |||
| BC004304.1 | - | 3 | 731 | 3625247 | ||
| Gene Symbol: LOC105375355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005250612.3 | XP_005250669.1 | 3 | 1228 | ||
| XM_006716140.3 | XP_006716203.1 | 5 | 900 | |||
| XM_006716141.3 | XP_006716204.1 | 4 | 820 | |||

UPK3B
Genomic Map