siRNA Details
| Gene Symbol: RAET1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130900.2 | NP_570970.2 | 3 | 494 | ||
| XM_011535486.2 | XP_011533788.1 | 3 | 558 | |||
| GenBank mRNA | AY039682.1 | - | 3 | 433 | ||
| BC131600.1 | - | 3 | 426 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAET1L| Gene Symbol: RAET1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130900.2 | NP_570970.2 | 3 | 494 | ||
| XM_011535486.2 | XP_011533788.1 | 3 | 558 | |||
| GenBank mRNA | AY039682.1 | - | 3 | 433 | ||
| BC131600.1 | - | 3 | 426 | |||