siRNA Details
| Gene Symbol: ZNF675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138330.2 | NP_612203.2 | 3 | 371 | ||
| XM_017026442.1 | XP_016881931.1 | 3 | 376 | |||
| XM_017026443.1 | XP_016881932.1 | 3 | 376 | |||
| XM_017026444.1 | XP_016881933.1 | 3 | 368 | |||
| GenBank mRNA | AK093669.1 | - | 3 | 358 | ||
| AK307845.1 | - | 3 | 299 | |||
| AY044432.1 | - | 3 | 296 | |||
| BC032394.1 | - | 3 | 353 | |||
| DA777369.1 | - | 3 | 390 | |||

ZNF675
Genomic Map