siRNA Details
| Gene Symbol: NME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002513.2 | NP_002504.2 | 2 | 323 | ||
| XM_005255332.4 | XP_005255389.1 | 2 | 354 | |||
| XM_011522503.2 | XP_011520805.1 | 2 | 364 | |||
| XM_011522504.2 | XP_011520806.1 | 2 | 359 | |||
| GenBank mRNA | AK308663.1 | - | 2 | 124 | ||
| BC000250.1 | - | 2 | 323 | 3357930 | ||
| BG911570.1 | - | 2 | 147 | |||
| U29656.1 | - | 2 | 166 | |||

NME3
Genomic Map