siRNA Details
| Gene Symbol: CHRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304472.1 | NP_001291401.1 | 3 | 554 | ||
| NM_001304473.1 | NP_001291402.1 | 4 | 609 | |||
| NM_001304474.1 | NP_001291403.1 | 3 | 554 | |||
| NM_003741.3 | NP_003732.2 | 3 | 554 | |||
| NR_130747.1 | 3 | 554 | ||||
| XM_011513254.1 | XP_011511556.1 | 3 | 407 | |||
| XM_011513256.1 | XP_011511558.1 | 2 | 195 | |||
| XM_017007388.1 | XP_016862877.1 | 3 | 407 | |||
| XM_017007389.1 | XP_016862878.1 | 3 | 407 | |||
| XM_017007390.1 | XP_016862879.1 | 3 | 407 | |||
| XM_017007391.1 | XP_016862880.1 | 3 | 407 | |||
| XM_017007392.1 | XP_016862881.1 | 3 | 407 | |||
| XM_017007393.1 | XP_016862882.1 | 3 | 407 | |||
| XM_017007394.1 | XP_016862883.1 | 2 | 195 | |||
| GenBank mRNA | AB208947.1 | - | 3 | 327 | ||
| AF209928.1 | - | 3 | 554 | |||
| AF209929.1 | - | 4 | 605 | |||
| AF209930.1 | - | 4 | 609 | |||
| AF283325.1 | - | 3 | 554 | |||
| AK074147.1 | - | 3 | 307 | |||
| AK293456.1 | - | 3 | 276 | |||
| AK295632.1 | - | 3 | 194 | |||
| AK296316.1 | - | 2 | 153 | |||
| AY358926.1 | - | 3 | 444 | |||
| BC112345.1 | - | 3 | 339 | 40008417 | ||

CHRD
Genomic Map