siRNA Details
| Gene Symbol: ERLIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003790.3 | NP_001003790.1 | 4 | 553 | ||
| NM_001003791.2 | NP_001003791.1 | 4 | 324 | |||
| NM_007175.6 | NP_009106.1 | 4 | 324 | |||
| XM_005273392.2 | XP_005273449.1 | 4 | 484 | |||
| XM_017013000.1 | XP_016868489.1 | 4 | 322 | |||
| GenBank mRNA | AK291394.1 | - | 4 | 277 | ||
| AK297279.1 | - | 4 | 277 | |||
| AL442077.1 | - | 311 | ||||
| AY358108.1 | - | 4 | 268 | |||
| AY358851.1 | - | 4 | 258 | |||
| BC005950.1 | - | 4 | 247 | |||
| BC048308.1 | - | 4 | 547 | |||
| BC050611.1 | - | 4 | 278 | |||
| BC067765.1 | - | 4 | 551 | 5296776 | ||
| BI560439.1 | - | 4 | 557 | |||
| BP353279.1 | - | 4 | 324 | |||

ERLIN2
Genomic Map