siRNA Details
| Gene Symbol: OR5V1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030876.5 | NP_110503.3 | 1 | 439 | ||
| XM_011514929.2 | XP_011513231.1 | 3 | 614 | |||
| XM_011514930.2 | XP_011513232.1 | 2 | 492 | |||
| GenBank mRNA | AJ459859.1 | - | 1 | 442 | ||
| BC069149.1 | - | 1 | 357 | |||

OR5V1
Genomic Map