siRNA Details
| Gene Symbol: UBALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC008681.1 | - | 1359 | |||
| BC012018.1 | - | 1326 | 4477559 | |||
| BC025327.1 | - | 1347 | 3614122 | |||
| BC062534.1 | - | 1404 | 4620605 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBALD1| Gene Symbol: UBALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC008681.1 | - | 1359 | |||
| BC012018.1 | - | 1326 | 4477559 | |||
| BC025327.1 | - | 1347 | 3614122 | |||
| BC062534.1 | - | 1404 | 4620605 | |||