siRNA Details
| Gene Symbol: IRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002199.3 | NP_002190.2 | 4 | 410 | ||
| GenBank mRNA | AK312953.1 | - | 4 | 388 | ||
| BC015803.2 | - | 3 | 217 | 3920890 | ||
| BP200998.1 | - | 4 | 410 | |||
| BT007264.1 | - | 3 | 210 | |||
| CR457077.1 | - | 3 | 210 | |||
| X15949.1 | - | 4 | 308 | |||

IRF2
Genomic Map