siRNA Details
| Gene Symbol: DOK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018431.4 | NP_060901.2 | 2 | 605 | ||
| NM_177959.2 | NP_808874.1 | 2 | 251 | |||
| XM_011528904.1 | XP_011527206.1 | 2 | 234 | |||
| GenBank mRNA | AF132732.1 | - | 2 | 327 | ||
| AF466368.1 | - | 2 | 130 | |||
| AK313222.1 | - | 2 | 317 | |||
| BC008992.1 | - | 2 | 251 | 4183000 | ||
| BI758267.1 | - | 2 | 350 | |||

DOK5
Genomic Map