siRNA Details
| Gene Symbol: ZNF626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145297.3 | NP_660340.1 | 4 | 418 | ||
| GenBank mRNA | BC007116.1 | - | 4 | 397 | ||
| BC107803.1 | - | 4 | 422 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF626| Gene Symbol: ZNF626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145297.3 | NP_660340.1 | 4 | 418 | ||
| GenBank mRNA | BC007116.1 | - | 4 | 397 | ||
| BC107803.1 | - | 4 | 422 | |||