siRNA Details
| Gene Symbol: ZNF493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145326.2 | NP_663299.2 | 4 | 1461 | ||
| GenBank mRNA | BC006408.1 | - | 4 | 1422 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF493| Gene Symbol: ZNF493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145326.2 | NP_663299.2 | 4 | 1461 | ||
| GenBank mRNA | BC006408.1 | - | 4 | 1422 | ||