siRNA Details
| Gene Symbol: NTN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145807.2 | NP_665806.1 | 7 | 1901 | ||
| GenBank mRNA | BC018654.2 | - | 7 | 1827 | 2961115 | |
| BC018697.2 | - | 7 | 1827 | 2961115 | ||
| BC033207.1 | - | 7 | 1827 | 4903484 | ||

NTN5
Genomic Map