siRNA Details
| Gene Symbol: SAMD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152486.2 | NP_689699.2 | 14 | 2255 | ||
| GenBank mRNA | BC024295.2 | - | 10 | 1830 | 3354502 | |
| BC033213.1 | - | 8 | 1620 | 5014368 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAMD11| Gene Symbol: SAMD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152486.2 | NP_689699.2 | 14 | 2255 | ||
| GenBank mRNA | BC024295.2 | - | 10 | 1830 | 3354502 | |
| BC033213.1 | - | 8 | 1620 | 5014368 | ||