siRNA Details
| Gene Symbol: ADCK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174922.4 | NP_777582.4 | 14 | 1700 | ||
| XM_006716527.2 | XP_006716590.1 | 14 | 1690 | |||
| XM_011516907.2 | XP_011515209.1 | 14 | 2413 | |||
| XM_011516909.1 | XP_011515211.1 | 14 | 1700 | |||
| XM_011516910.1 | XP_011515212.1 | 15 | 1723 | |||
| XM_017013174.1 | XP_016868663.1 | 14 | 1738 | |||
| GenBank mRNA | AK092773.1 | - | 14 | 1663 | ||
| BC031570.1 | - | 3 | 433 | |||
| BC064345.1 | - | 5 | 651 | |||
| BC085013.1 | - | 9 | 1010 | |||
| BC101659.1 | - | 14 | 1640 | 8069165 | ||
| BC143326.1 | - | 13 | 1467 | |||
| BI825038.1 | - | 3 | 463 | |||

ADCK5
Genomic Map