siRNA Details
| Gene Symbol: UBR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175748.3 | NP_786924.2 | 1 | 376 | ||
| NR_038150.1 | 1 | 376 | ||||
| GenBank mRNA | AK097477.1 | - | 1 | 361 | ||
| BC015046.1 | - | 1 | 175 | 3908032 | ||
| BC017013.1 | - | 1 | 154 | 3883770 | ||
| BC051819.1 | - | 1 | 138 | 6171393 | ||
| BX248249.1 | - | 1 | 135 | |||
| DA158479.1 | - | 1 | 150 | |||
| DB047433.1 | - | 1 | 376 | |||

UBR7
Genomic Map