siRNA Details
Gene Symbol: LINC00342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057596.1 | - | 1 | 89 | ||
BC117383.1 | - | 1 | 59 | |||
BC126153.1 | - | 1 | 59 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057596.1 | - | 1 | 89 | ||
BC117383.1 | - | 1 | 59 | |||
BC126153.1 | - | 1 | 59 |