siRNA Details
| Gene Symbol: TAS2R31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176885.2 | NP_795366.2 | 1 | 182 | ||
| GenBank mRNA | BC117421.1 | - | 1 | 183 | 40125972 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R31| Gene Symbol: TAS2R31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176885.2 | NP_795366.2 | 1 | 182 | ||
| GenBank mRNA | BC117421.1 | - | 1 | 183 | 40125972 | |