siRNA Details
| Gene Symbol: NKX6-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177400.2 | NP_796374.1 | 2 | 525 | ||
| XM_017016789.1 | XP_016872278.1 | 2 | 519 | |||
| GenBank mRNA | BC101635.1 | - | 2 | 525 | 8069141 | |
| BC104823.1 | - | 2 | 525 | 8143826 | ||
| BF509028.1 | - | 2 | 155 | |||

NKX6-2
Genomic Map