siRNA Details
| Gene Symbol: VN1R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173857.2 | NP_776256.2 | 1 | 149 | ||
| XM_011526885.1 | XP_011525187.1 | 2 | 301 | |||
| GenBank mRNA | BC104935.1 | - | 1 | 203 | 8143938 | |
| BC104939.1 | - | 1 | 207 | 8143942 | ||

VN1R4
Genomic Map