siRNA Details
Gene Symbol: VN1R5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173858.1 | NP_776257.1 | 1 | 493 | ||
GenBank mRNA | BC126341.1 | - | 1 | 593 | 8992057 | |
BC126367.1 | - | 1 | 593 | 8992083 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VN1R5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173858.1 | NP_776257.1 | 1 | 493 | ||
GenBank mRNA | BC126341.1 | - | 1 | 593 | 8992057 | |
BC126367.1 | - | 1 | 593 | 8992083 |