siRNA Details
Gene Symbol: NLRP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321057.1 | NP_001307986.1 | 5 | 815 | ||
NM_176810.2 | NP_789780.2 | 5 | 815 | |||
GenBank mRNA | AY154468.1 | - | 5 | 790 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NLRP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321057.1 | NP_001307986.1 | 5 | 815 | ||
NM_176810.2 | NP_789780.2 | 5 | 815 | |||
GenBank mRNA | AY154468.1 | - | 5 | 790 |