siRNA Details
| Gene Symbol: LYVE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006691.3 | NP_006682.2 | 1 | 361 | ||
| GenBank mRNA | AF118108.1 | - | 1 | 165 | ||
| AF127670.2 | - | 1 | 195 | |||
| AK075443.1 | - | 1 | 275 | |||
| AK300885.1 | - | 1 | 120 | |||
| AK312462.1 | - | 1 | 359 | |||
| AY358925.1 | - | 1 | 234 | |||
| BC026231.1 | - | 1 | 120 | 4712863 | ||
| DA924658.1 | - | 1 | 361 | |||

LYVE1
Genomic Map