siRNA Details
| Gene Symbol: NLGN4Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206850.1 | NP_001193779.1 | 5 | 887 | ||
| NM_014893.4 | NP_055708.3 | 5 | 1217 | |||
| NR_028319.1 | 5 | 1439 | ||||
| NR_046355.1 | 6 | 1204 | ||||
| XM_006724874.2 | XP_006724937.1 | 6 | 1449 | |||
| XM_011531424.2 | XP_011529726.1 | 6 | 1767 | |||
| XM_011531425.2 | XP_011529727.1 | 6 | 1539 | |||
| XM_011531426.2 | XP_011529728.1 | 6 | 1500 | |||
| XM_011531427.1 | XP_011529729.1 | 6 | 1254 | |||
| XM_011531428.1 | XP_011529730.1 | 7 | 1435 | |||
| XM_011531429.2 | XP_011529731.1 | 6 | 2050 | |||
| XM_011531430.2 | XP_011529732.1 | 6 | 1388 | |||
| XM_017030034.1 | XP_016885523.1 | 5 | 1479 | |||
| XM_017030035.1 | XP_016885524.1 | 5 | 1400 | |||
| XM_017030036.1 | XP_016885525.1 | 5 | 1194 | |||
| XM_017030037.1 | XP_016885526.1 | 5 | 1192 | |||
| XM_017030038.1 | XP_016885527.1 | 5 | 1990 | |||
| XM_017030039.1 | XP_016885528.1 | 6 | 725 | |||
| XM_017030040.1 | XP_016885529.1 | 5 | 574 | |||
| GenBank mRNA | AB023168.1 | - | 5 | 743 | ||
| AF376804.1 | - | 5 | 1217 | |||
| AK295117.1 | - | 6 | 1204 | |||
| BC113525.1 | - | 4 | 1044 | 8322577 | ||
| BC113551.1 | - | 4 | 1044 | 8322603 | ||
| BX537428.1 | - | 5 | 1440 | |||

NLGN4Y
Genomic Map