siRNA Details
| Gene Symbol: TRPV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016113.4 | NP_057197.2 | 2 | 487 | ||
| XM_005256676.1 | XP_005256733.1 | 2 | 487 | |||
| XM_005256677.1 | XP_005256734.1 | 2 | 487 | |||
| XM_005256678.4 | XP_005256735.1 | 2 | 487 | |||
| XM_006721541.3 | XP_006721604.1 | 2 | 487 | |||
| XM_006721543.3 | XP_006721606.1 | 2 | 487 | |||
| XM_011523922.1 | XP_011522224.1 | 2 | 487 | |||
| XM_011523923.1 | XP_011522225.1 | 2 | 487 | |||
| XM_017024730.1 | XP_016880219.1 | 2 | 487 | |||
| XM_017024731.1 | XP_016880220.1 | 2 | 487 | |||
| XM_017024732.1 | XP_016880221.1 | 2 | 487 | |||
| GenBank mRNA | AF103906.1 | - | 2 | 253 | ||
| AF129112.1 | - | 1 | 123 | |||
| AJ487963.1 | - | 1 | 92 | |||
| AK001896.1 | - | 2 | 417 | |||
| AK289705.1 | - | 2 | 487 | |||
| BC018926.2 | - | 2 | 455 | 4298484 | ||
| BC051305.1 | - | 2 | 443 | 4870010 | ||

TRPV2
Genomic Map