siRNA Details
| Gene Symbol: POU2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198783.1 | NP_001185712.1 | 4 | 465 | ||
| NM_001198786.1 | NP_001185715.1 | 4 | 332 | |||
| NM_002697.3 | NP_002688.3 | 4 | 332 | |||
| NR_037163.1 | 5 | 376 | ||||
| XM_011509653.2 | XP_011507955.1 | 5 | 284 | |||
| XM_011509654.2 | XP_011507956.1 | 4 | 245 | |||
| XM_011509655.1 | XP_011507957.1 | 5 | 284 | |||
| XM_017001508.1 | XP_016856997.1 | 5 | 284 | |||
| GenBank mRNA | AK091438.1 | - | 6 | 760 | ||
| AK302525.1 | - | 5 | 371 | |||
| AK303201.1 | - | 4 | 270 | |||
| AK308004.1 | - | 5 | 371 | |||
| AY113189.1 | - | 4 | 284 | |||
| BC001664.2 | - | 4 | 248 | 2966289 | ||
| BC003571.1 | - | 4 | 248 | 3544378 | ||
| BC052274.1 | - | 5 | 356 | 4622256 | ||
| BX954888.1 | - | 4 | 333 | |||
| DA937884.1 | - | 4 | 465 | |||
| DC370634.1 | - | 5 | 376 | |||
| KT438684.1 | - | 6 | 443 | |||
| X13403.1 | - | 4 | 253 | |||

POU2F1
Genomic Map