siRNA Details
| Gene Symbol: WWC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015691.3 | NP_056506.2 | 7 | 725 | ||
| GenBank mRNA | AB033106.1 | - | 5 | 348 | ||
| AK292961.1 | - | 6 | 607 | |||
| AL353937.1 | - | 3 | 205 | |||
| BC032447.1 | - | 4 | 260 | |||
| KC987947.1 | - | 7 | 902 | |||

WWC3
Genomic Map