siRNA Details
| Gene Symbol: PRODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016335.4 | NP_057419.4 | 2, 3 | 485 | ||
| GenBank mRNA | AB209472.1 | - | 1, 2 | 136 | ||
| BC036534.1 | - | 1, 2 | 294 | |||
| BC068260.1 | - | 1, 2 | 287 | |||
| CN362765.1 | - | 1, 2 | 236 | |||
| KJ534920.1 | - | 1, 2 | 281 | |||
| U79754.1 | - | 2, 3 | 485 | |||
| Gene Symbol: LOC102724788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724935.3 | XP_006724998.1 | 1, 2 | 314 | ||
| XM_006724936.3 | XP_006724999.1 | 1, 2 | 289 | |||

PRODH
Genomic Map