siRNA Details
| Gene Symbol: ZC3H7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017590.5 | NP_060060.3 | 3, 4 | 360 | ||
| XM_011530016.2 | XP_011528318.1 | 3, 4 | 341 | |||
| GenBank mRNA | AK023849.1 | - | 3, 4 | 148 | ||
| AK225891.1 | - | 3, 4 | 333 | |||
| AK315009.1 | - | 3, 4 | 302 | |||
| BC098378.1 | - | 3, 4 | 298 | |||
| BC152558.1 | - | 3, 4 | 338 | 8860405 | ||

ZC3H7B
Genomic Map