siRNA Details
| Gene Symbol: WWC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015691.3 | NP_056506.2 | 8 | 904 | ||
| GenBank mRNA | AB033106.1 | - | 6 | 527 | ||
| AK292961.1 | - | 7 | 786 | |||
| AK301129.1 | - | 1 | 167 | |||
| AL353937.1 | - | 4 | 384 | |||
| BC032447.1 | - | 5 | 439 | |||
| KC987947.1 | - | 8 | 1081 | |||

WWC3
Genomic Map